1-Chloro-3-fluorobenzene ≥99%

Supplier: Matrix Scientific

002346-100G 002346-25G
101775-992EA 86.15 USD
101775-992 101775-990
1-Chloro-3-fluorobenzene ≥99%
1-Chloro-3-fluorobenzene

TCA listed: T
Hazard: Flammable

Formula: C₆H₄ClF
Boiling Pt: 126-128°C
Density: 1.219
MDL Number: MFCD00000569
CAS Number: 625-98-9

Order Now

Specification Test Results

Boiling point 126-128°
Density 1.219
ND20 1.494


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR