2-Chloro-5-(trifluoromethyl)benzonitrile ≥98%

Supplier: Matrix Scientific

002339-25G 002339-100G 002339-250G
101775-984EA 58.46 USD
101775-984 101775-986 101775-988
2-Chloro-5-(trifluoromethyl)benzonitrile ≥98%
2-Chloro-5-(trifluoromethyl)benzonitrile

TCA listed: T
Hazard: Toxic/Irritant

Formula: C₈H₃ClF₃N
Boiling Pt: 108…109 °C (10 mmHg)
Melting Pt: 35…37 °C
MDL Number: MFCD00019742
CAS Number: 328-87-0
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 108-109°/1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR