2-Chloro-3-(trifluoromethyl)benzyl bromide

Supplier: Matrix Scientific

002308-1G 002308-5G
101775-970EA 49.23 USD
101775-970 101775-972
2-Chloro-3-(trifluoromethyl)benzyl bromide

Hazard: Corrosive/Lachrymator

Formula: C₈H₅BrClF₃
MDL Number: MFCD01631539
CAS Number: 261763-22-8
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR