4-Fluoro-3-(trifluoromethyl)phenol ≥98%

Supplier: Matrix Scientific

002302-25G 002302-5G
101775-964EA 121.54 USD
101775-964 101775-962
4-Fluoro-3-(trifluoromethyl)phenol ≥98%
4-Fluoro-3-(trifluoromethyl)phenol

Hazard: Corrosive/Irritant

Formula: C₇H₄F₄O
MDL Number: MFCD00043875
CAS Number: 61721-07-1
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR