4-Chloro-3-(trifluoromethyl)benzylamine

Supplier: Matrix Scientific

002298-5G 002298-25G
101775-954EA 36.92 USD
101775-954 101775-956
4-Chloro-3-(trifluoromethyl)benzylamine
4-Chloro-3-(trifluoromethyl)benzylamine

Hazard: Corrosive

Formula: C₈H₇ClF₃N
MDL Number: MFCD00190124
CAS Number: 62039-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR