2-Fluoro-3-(trifluoromethyl)phenol ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-3-hydroxybenzotrifluoride

002290-1G 002290-5G
101775-942EA 44.62 USD
101775-942 101775-944
2-Fluoro-3-(trifluoromethyl)phenol ≥98%
2-Fluoro-3-(trifluoromethyl)phenol

Hazard: Toxic

Formula: C₇H₄F₄O
MW: 180.1 g/mol
MDL Number: MFCD00083554
CAS Number: 207291-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR