4-Fluoro-3-(trifluoromethyl)benzyl bromide ≥98%

Supplier: Matrix Scientific

Synonyms: 4-(Bromomethyl)-1-fluoro-2-(trifluoromethyl)benzene, 1-(Bromomethyl)-4-fluoro-3-(trifluoromethyl)benzene

002289-5G 002289-25G 002289-1G
101775-938EA 41.54 USD
101775-938 101775-940 101775-936
4-Fluoro-3-(trifluoromethyl)benzyl bromide ≥98%
4-Fluoro-3-(trifluoromethyl)benzyl bromide

Hazard: Corrosive/Refrigerate/Lachrymator

Formula: C₈H₅BrF₄
MW: 257.03 g/mol
MDL Number: MFCD00061177
CAS Number: 184970-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR