4-Fluoro-3-(trifluoromethyl)benzoyl chloride ≥97%

Supplier: Matrix Scientific

002284-1G 002284-5G
101775-924EA 18.46 USD
101775-924 101775-926
4-Fluoro-3-(trifluoromethyl)benzoyl chloride ≥97%
4-Fluoro-3-(trifluoromethyl)benzoyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₈H₃ClF₄O
MW: 226.56 g/mol
MDL Number: MFCD00061159
CAS Number: 67515-56-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR