2-Chloro-3,6-difluorophenol

Supplier: Matrix Scientific

Synonyms: 2-Choro-3,6-difluorophenol

002269-1G 002269-5G
101775-910EA 53.85 USD
101775-910 101775-912
2-Chloro-3,6-difluorophenol
2-Chloro-3,6-difluorophenol

Hazard: Corrosive

Formula: C₆H₃ClF₂O
MW: 164.54 g/mol
Melting Pt: 52…56 °C
MDL Number: MFCD01631450
CAS Number: 261762-50-9
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR