α-Bromo-2-chloro-3,6-difluorotoluene

Supplier: Matrix Scientific

002267-1G 002267-5G
101775-906EA 49.23 USD
101775-906 101775-908
α-Bromo-2-chloro-3,6-difluorotoluene
α-Bromo-2-chloro-3,6-difluorotoluene

Hazard: Corrosive/Refrigerate/Lachrymator

Formula: C₇H₄BrClF₂
MDL Number: MFCD01631446
CAS Number: 90292-67-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR