2-Chloro-3,6-difluorobenzylamine

Supplier: Matrix Scientific

002266-1G 002266-5G
101775-902EA 47.69 USD
101775-902 101775-904
2-Chloro-3,6-difluorobenzylamine
2-Chloro-3,6-difluorobenzylamine

Hazard: Corrosive

Formula: C₇H₆ClF₂N
MDL Number: MFCD01631449
CAS Number: 261762-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR