2-Chloro-3,6-difluorobenzoyl chloride

Supplier: Matrix Scientific

Synonyms: 2-Chloro-3,6-difluorobenzene-1-carbonyl chloride

002264-1G 002264-5G
101775-898EA 36.92 USD
101775-898 101775-900
2-Chloro-3,6-difluorobenzoyl chloride
2-Chloro-3,6-difluorobenzoyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₇H₂Cl₂F₂O
MDL Number: MFCD01631442
CAS Number: 261762-42-9
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

ND20 1.51

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR