4-Chloro-1,2-difluorobenzene ≥97%

Supplier: Matrix Scientific

Synonyms: 3,4-Difluorochlorobenzene

002253-25G 002253-100G
101775-890EA 73.85 USD
101775-890 101775-892
4-Chloro-1,2-difluorobenzene ≥97%
4-Chloro-1,2-difluorobenzene

Hazard: Flammable

Formula: C₆H₃ClF₂
MW: 148.54 g/mol
Boiling Pt: 127°C
MDL Number: MFCD00042572
CAS Number: 696-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR