α-Methyl-2-trifluoromethylbenzyl bromide ≥97%

Supplier: Matrix Scientific

002212-1G 002212-5G
101775-862EA 123.08 USD
101775-862 101775-864
α-Methyl-2-trifluoromethylbenzyl bromide ≥97%
alpha-Methyl-2-trifluoromethylbenzyl bromide

Hazard: Corrosive/Lachrymator

Formula: C₉H₈BrF₃
MDL Number: MFCD00039226
CAS Number: 194152-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR