Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent) ≥95%
Supplier: Matrix Scientific
Synonyms:
Diethyl [(ethoxycarbonyl)(fluoro)methyl]phosphonate, Triethyl 2-fluoro-2-phosphonoacetate
002202-5G
002202-25G
101775-858EA
73.85
USD
101775-858
101775-860
Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent) ≥95%
Triethyl 2-fluoro-2-phosphonoacetate (fluorinated Horner reagent)
Hazard: Flammable/Irritant
Formula:
C₈H₁₆FO₅P MW: 242.19 g/mol Boiling Pt: 75 °C (0.01 mmHg) Density: 1.194 |
MDL Number:
MFCD00134400 CAS Number: 2356-16-3 |
Specification Test Results
Boiling point | 75°/0.01mm |
Density | 1.194 |
ND20 | 1.425 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...