2-Methyl-2-pentenoic acid ≥98%
Supplier: Matrix Scientific
002201-100G
002201-25G
101775-856EA
136.92
USD
101775-856
101775-854
2-Methyl-2-pentenoic acid ≥98%
2-Methyl-2-pentenoic acid
TCA listed: T
Hazard: Corrosive/Stench
Formula:
C₆H₁₀O₂ Boiling Pt: 120 °C (20 mmHg) Density: 0.978 Flash Pt: 107 °C |
MDL Number:
MFCD00002655 CAS Number: 3142-72-1 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...