5-Fluoro-2-methylphenylacetonitrile ≥97%
Supplier: Matrix Scientific
Synonyms:
2-(5-Fluoro-2-methylphenyl)acetonitrile
002182-1G
002182-5G
002182-25G
101775-840EA
43.08
USD
101775-840
101775-842
101775-844
5-Fluoro-2-methylphenylacetonitrile ≥97%
5-Fluoro-2-methylphenylacetonitrile
Hazard: Toxic
Formula:
C9H8FN MW: 149.1700 g/mol |
MDL Number:
MFCD01631529 CAS Number: 80141-97-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...