5-Fluoro-o-tolylisothiocyanate ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-5-fluorophenylisothiocyanate, 5-Fluoro-2-methylphenyl Isothiocyanate, Isothiocyanic Acid 5-Fluoro-o-tolyl Ester

002179-5G 002179-1G
101775-838EA 92.31 USD
101775-838 101775-836
5-Fluoro-o-tolylisothiocyanate ≥97%
5-Fluoro-o-tolylisothiocyanate

Hazard: Irritant

Formula: C₈H₆FNS
MW: 167.20 g/mol
Boiling Pt: 101 °C (20 mmHg)
MDL Number: MFCD00046800
CAS Number: 175205-39-7

Order Now

Specification Test Results

Boiling point 101°/20mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR