5-Fluoro-2-methylbenzyl bromide

Supplier: Matrix Scientific

Synonyms: 2-(Bromomethyl)-4-fluoro-1-methylbenzene

002166-1G 002166-5G
101775-828EA 35.38 USD
101775-828 101775-830
5-Fluoro-2-methylbenzyl bromide
5-Fluoro-2-methylbenzyl bromide

Hazard: Corrosive

Formula: C₈H₈BrF
MW: 203.05 g/mol
MDL Number: MFCD01631528
CAS Number: 261951-71-7
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR