5-Fluoro-2-toluoyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Fluoro-2-methylbenzene-1-carbonyl chloride, 5-Fluoro-2-methylbenzoyl chloride

002161-1G 002161-5G
101775-820EA 26.15 USD
101775-820 101775-822
5-Fluoro-2-toluoyl chloride ≥97%
5-Fluoro-2-toluoyl chloride

Hazard: Corrosive/Refrigerate

Formula: C₈H₆ClFO
MDL Number: MFCD00236681
CAS Number: 21900-39-0
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR