4-Fluoro-o-toluidine ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-fluorotoluene, 4-Fluoro-2-methylaniline

002158-25G 002158-100G
101775-816EA 18.46 USD
101775-816 101775-818
4-Fluoro-o-toluidine ≥98%
4-Fluoro-o-toluidine

TCA listed: T
Hazard: Toxic/Irritant

Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 207
Melting Pt: 14
Density: 1.126
MDL Number: MFCD00007832
CAS Number: 452-71-1
UN: 2810
ADR: 6.1 III

Order Now

Specification Test Results

Boiling point 90-92°/16m
Density 1.126
ND20 1.537


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR