3-Fluoro-o-toluidine ≥99%

Supplier: Matrix Scientific

Synonyms: 2-Amino-6-fluorotoluene, 3-Fluoro-2-methylaniline

002157-25G 002157-100G
101775-814EA 63.08 USD
101775-814 102550-816
3-Fluoro-o-toluidine ≥99%
3-Fluoro-o-toluidine

Hazard: Toxic/Irritant

Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 89-91°C/15mm
Melting Pt: 7°C
Density: 1.099
MDL Number: MFCD00007760
CAS Number: 443-86-7
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 89-91°/15m
Density 1.099
ND20 1.542

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR