1-Fluoro-2-iodoethane ≥98% stabilised

Supplier: Matrix Scientific


Stabilization: Stabilised with copper 1%

002137-1G 002137-5G
101775-804EA 58.46 USD
101775-804 101775-806
1-Fluoro-2-iodoethane ≥98% stabilised
1-Fluoro-2-iodoethane

contains up to 1% copper powder as stabilizer


Hazard: Toxic/Refrigerate

Formula: C₂H₄FI
Boiling Pt: 98-102°C
MDL Number: MFCD00748131
CAS Number: 762-51-6

Order Now

Specification Test Results

Boiling point 98-102°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR