5-Chloro-2-fluorotoluene ≥99%

Supplier: Matrix Scientific

002117-100G 002117-25G
101775-800EA 143.08 USD
101775-800 101775-798
5-Chloro-2-fluorotoluene ≥99%
5-Chloro-2-fluorotoluene

Hazard: Flammable

Formula: C₇H₆ClF
Boiling Pt: 156 °C
Density: 1.19
MDL Number: MFCD00018504
CAS Number: 452-66-4

Order Now

Specification Test Results

Boiling point 156°
Density 1.19
ND20 1.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR