5-Chloro-2-fluorobenzylamine

Supplier: Matrix Scientific

002099-5G 002099-1G
101775-774EA 90.77 USD
101775-774 101775-772
5-Chloro-2-fluorobenzylamine
5-Chloro-2-fluorobenzylamine

Hazard: Corrosive

Formula: C₇H₇ClFN
MW: 159.59 g/mol
MDL Number: MFCD01631440
CAS Number: 261723-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR