5-Chloro-2-fluorobenzoyl chloride ≥98%

Supplier: Matrix Scientific

002095-5G 002095-25G
101775-762EA 27.69 USD
101775-762 101775-764
5-Chloro-2-fluorobenzoyl chloride ≥98%
5-Chloro-2-fluorobenzoyl chloride

Hazard: Corrosive/Irritant

Formula: C₇H₃Cl₂FO
Boiling Pt: 103-104°C/15mm
MDL Number: MFCD01631433
CAS Number: 394-29-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 103-104°/1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR