5-Chloro-2-fluorobenzamide
Supplier: Matrix Scientific
002092-1G
002092-5G
002092-25G
101775-752EA
23.08
USD
101775-752
101775-754
101775-756
5-Chloro-2-fluorobenzamide
5-Chloro-2-fluorobenzamide
Hazard: Irritant
Formula:
C₇H₅ClFNO |
MDL Number:
MFCD01631435 CAS Number: 261762-57-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...