5-Chloro-2-fluoroaniline ≥97%

Supplier: Matrix Scientific

002084-100G 002084-25G
101775-750EA 96.92 USD
101775-750 101775-748
5-Chloro-2-fluoroaniline ≥97%
5-Chloro-2-fluoroaniline

Hazard: Toxic

Formula: C₆H₅ClFN
Boiling Pt: 102…105 °C (20 mmHg)
Density: 1.3
MDL Number: MFCD00069416
CAS Number: 2106-05-0
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR