3-Chloro-2-fluoro-5-(trifluoromethyl)phenol

Supplier: Matrix Scientific

002069-1G 002069-5G
101775-740EA 60 USD
101775-740 101775-742
3-Chloro-2-fluoro-5-(trifluoromethyl)phenol
3-Chloro-2-fluoro-5-(trifluoromethyl)phenol

Hazard: Corrosive/Irritant

Formula: C₇H₃ClF₄O
MW: 214.55 g/mol
MDL Number: MFCD01631514
CAS Number: 261763-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR