3-Chloro-2-fluoro-5-(trifluoromethyl)benzylbromide

Supplier: Matrix Scientific

002067-1G 002067-5G
101775-736EA 89.23 USD
101775-736 101775-738
3-Chloro-2-fluoro-5-(trifluoromethyl)benzylbromide
3-Chloro-2-fluoro-5-(trifluoromethyl)benzylbromide

Hazard: Corrosive/Lachrymator

Formula: C₈H₄BrClF₄
MDL Number: MFCD01631513
CAS Number: 261763-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR