3-Chloro-2-fluoro-5-(trifluoromethyl)benzoyl chloride

Supplier: Matrix Scientific

002064-1G 002064-5G
101775-732EA 32.31 USD
101775-732 101775-734
3-Chloro-2-fluoro-5-(trifluoromethyl)benzoyl chloride
3-Chloro-2-fluoro-5-(trifluoromethyl)benzoyl chloride

Hazard: Corrosive

Formula: C₈H₂Cl₂F₄O
MW: 261 g/mol
MDL Number: MFCD01631511
CAS Number: 261763-03-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR