6-Chloro-2-fluoro-m-cresol

Supplier: Matrix Scientific

002056-1G 002056-5G
101775-724EA 55.38 USD
101775-724 101775-726
6-Chloro-2-fluoro-m-cresol
6-Chloro-2-fluoro-m-cresol

Hazard: Corrosive

Formula: C₇H₆ClFO
Melting Pt: 46…49 °C
MDL Number: MFCD01631376
CAS Number: 261762-91-8
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR