6-Chloro-2-fluoro-3-methylbenzyl bromide

Supplier: Matrix Scientific

Synonyms: 2-(Bromomethyl)-1-chloro-3-fluoro-4-methylbenzene

002055-1G 002055-5G
101775-720EA 61.54 USD
101775-720 101775-722
6-Chloro-2-fluoro-3-methylbenzyl bromide
6-Chloro-2-fluoro-3-methylbenzyl bromide

Hazard: Corrosive/Lachrymator

Formula: C₈H₇BrClF
MW: 237.51 g/mol
MDL Number: MFCD01631373
CAS Number: 261762-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR