6-Chloro-2-fluoro-3-methylbenzylamine

Supplier: Matrix Scientific

002054-1G 002054-5G
101775-716EA 78.46 USD
101775-716 101775-718
6-Chloro-2-fluoro-3-methylbenzylamine
6-Chloro-2-fluoro-3-methylbenzylamine

Hazard: Corrosive

Formula: C₈H₉ClFN
MDL Number: MFCD01631420
CAS Number: 261762-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR