6-Chloro-2-fluoro-3-methylbenzaldehyde

Supplier: Matrix Scientific

002049-1G 002049-5G
101775-708EA 40 USD
101775-708 101775-710
6-Chloro-2-fluoro-3-methylbenzaldehyde
6-Chloro-2-fluoro-3-methylbenzaldehyde

Hazard: Irritant

Formula: C₈H₆ClFO
MDL Number: MFCD01631370
CAS Number: 286474-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR