3-Fluoro-2-butanone

Supplier: Matrix Scientific

002044-1G 002044-5G
101775-704EA 226.15 USD
101775-704 101775-706
3-Fluoro-2-butanone
3-Fluoro-2-butanone

Hazard: Flammable

Formula: C₄H₇FO
MW: 90.1 g/mol
Boiling Pt: 74…76 °C
MDL Number: MFCD01862088
CAS Number: 814-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR