5-Fluoro-2-(trifluoromethyl)phenylacetonitrile

Supplier: Matrix Scientific

002015-5G 002015-1G
101775-684EA 166.15 USD
101775-684 101775-682
5-Fluoro-2-(trifluoromethyl)phenylacetonitrile
5-Fluoro-2-(trifluoromethyl)phenylacetonitrile

Hazard: Toxic

Formula: C₉H₅F₄N
MDL Number: MFCD06247720
CAS Number: 242812-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR