3-Fluoro-2-(trifluoromethyl)benzoyl chloride

Supplier: Matrix Scientific

002011-5G 002011-1G
101775-678EA 964.62 USD
101775-678 101775-676
3-Fluoro-2-(trifluoromethyl)benzoyl chloride
3-Fluoro-2-(trifluoromethyl)benzoyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₈H₃ClF₄O
MDL Number: MFCD01631458
CAS Number: 261951-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR