5-Chloro-2-(trifluoromethyl)phenylacetonitrile

Supplier: Matrix Scientific

002007-1G 002007-5G
101775-668EA 100 USD
101775-668 101775-670
5-Chloro-2-(trifluoromethyl)phenylacetonitrile
5-Chloro-2-(trifluoromethyl)phenylacetonitrile

Hazard: Toxic

Formula: C₉H₅ClF₃N
MDL Number: MFCD01631357
CAS Number: 261763-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR