5-Chloro-2-(trifluoromethyl)benzyl bromide

Supplier: Matrix Scientific

002005-1G 002005-5G
101775-664EA 30.77 USD
101775-664 101775-666
5-Chloro-2-(trifluoromethyl)benzyl bromide

Hazard: Corrosive/Lachrymator

Formula: C₈H₅BrClF₃
MDL Number: MFCD01631356
CAS Number: 261763-24-0
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR