5-Fluoro-2-(trifluoromethyl)benzyl bromide ≥98%

Supplier: Matrix Scientific

001996-5G 001996-25G
101775-656EA 41.54 USD
101775-656 101775-658
5-Fluoro-2-(trifluoromethyl)benzyl bromide ≥98%
5-Fluoro-2-(trifluoromethyl)benzyl bromide

Hazard: Corrosive/Lachrymator

Formula: C₈H₅BrF₄
MDL Number: MFCD00236290
CAS Number: 239135-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR