5-Fluoro-2-(trifluoromethyl)benzoyl chloride ≥95%
Supplier: Matrix Scientific
Synonyms:
alpha,alpha,alpha,5-Tetrafluoro-o-toluoyl chloride
001993-100G
001993-5G
001993-25G
101775-654EA
555.38
USD
101775-654
101775-650
101775-652
5-Fluoro-2-(trifluoromethyl)benzoyl chloride ≥95%
5-Fluoro-2-(trifluoromethyl)benzoyl chloride
Hazard: Corrosive/Moisture Sensitive/Lachrymator
Formula:
C₈H₃ClF₄O Boiling Pt: 168…169 °C Density: 1.5 |
MDL Number:
MFCD00083534 CAS Number: 216144-70-6 UN: 3265 ADR: 8,II |
Specification Test Results
Boiling point | 168-169° |
Density | 1.5 |
ND20 | 1.463 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...