5-Fluoro-2-(trifluoromethyl)benzonitrile ≥97%

Supplier: Matrix Scientific

001991-5G 001991-25G
101775-646EA 120 USD
101775-646 101775-648
5-Fluoro-2-(trifluoromethyl)benzonitrile ≥97%
5-Fluoro-2-(trifluoromethyl)benzonitrile

Hazard: Toxic/Irritant

Formula: C₈H₃F₄N
MW: 189.11 g/mol
MDL Number: MFCD00236281
CAS Number: 240800-45-7
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR