4-Fluoro-2-(trifluoromethyl)benzonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Cyano-5-fluorobenzotrifluoride

001977-25G 001977-100G
101775-638EA 26.15 USD
101775-638 101775-640
4-Fluoro-2-(trifluoromethyl)benzonitrile ≥97%
4-Fluoro-2-(trifluoromethyl)benzonitrile

Hazard: Toxic/Irritant

Formula: C₈H₃F₄N
MW: 189.11 g/mol
Boiling Pt: 52…55 °C (15 mmHg)
Melting Pt: 43…44 °C
Density: 1.35
MDL Number: MFCD00061283
CAS Number: 194853-86-6
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 52-55°/15m
Density 1.35

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR