α-Bromo-3-chloro-2,6-difluorotoluene

Supplier: Matrix Scientific

001970-1G 001970-5G
101775-634EA 53.85 USD
101775-634 101775-636
α-Bromo-3-chloro-2,6-difluorotoluene
α-Bromo-3-chloro-2,6-difluorotoluene

Hazard: Corrosive/Lachrymator

Formula: C₇H₄BrClF₂
MDL Number: MFCD01631326
CAS Number: 261762-47-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR