3-Chloro-2,6-difluorobenzylamine

Supplier: Matrix Scientific

001969-1G 001969-5G
101775-630EA 126.15 USD
101775-630 101775-632
3-Chloro-2,6-difluorobenzylamine
3-Chloro-2,6-difluorobenzylamine

TCA listed: F
Hazard: Corrosive/Irritant

Formula: C₇H₆ClF₂N
MDL Number: MFCD01631388
CAS Number: 261762-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR