3-Chloro-2,6-difluorobenzoyl chloride

Supplier: Matrix Scientific

001967-1G 001967-5G
101775-626EA 40 USD
101775-626 101775-628
3-Chloro-2,6-difluorobenzoyl chloride
3-Chloro-2,6-difluorobenzoyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₇H₂Cl₂F₂O
MDL Number: MFCD01631324
CAS Number: 261762-43-0
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

ND20 1.52


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR