2-Chloro-1,4-difluorobenzene ≥98%

Supplier: Matrix Scientific

Synonyms: 2,5-Difluorochlorobenzene

001957-25G 001957-100G
101775-624EA 84.62 USD
101775-624 102550-806
2-Chloro-1,4-difluorobenzene ≥98%
2-Chloro-1,4-difluorobenzene

Hazard: Flammable

Formula: C₆H₃ClF₂
MW: 148.54 g/mol
Boiling Pt: 128
Melting Pt: -24.6
Density: 1.352
Flash Pt: 20 °C
MDL Number: MFCD00042571
CAS Number: 2367-91-1

Order Now

Specification Test Results

Boiling point 126-128°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR