2-Chloro-1,1,2-trifluoroethyl ethyl ether ≥97%

Supplier: Matrix Scientific

001890-5G 001890-25G
101775-584EA 30.77 USD
101775-584 101775-586
2-Chloro-1,1,2-trifluoroethyl ethyl ether ≥97%
2-Chloro-1,1,2-trifluoroethyl ethyl ether

Hazard: Flammable

Formula: C₄H₆ClF₃O
MW: 162.54 g/mol
Boiling Pt: 82°C/630mm
Density: 1.273
MDL Number: MFCD00018849
CAS Number: 310-71-4

Order Now

Specification Test Results

Boiling point 82°/630mm
Density 1.273
ND20 1.348

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR