1,1,1,2,2-Pentafluoro-4-iodobutane ≥97% stabilized

Supplier: Matrix Scientific


Stabilization: Stabilised with copper

001887-5G 001887-25G
101775-580EA 41.54 USD
101775-580 101775-582
1,1,1,2,2-Pentafluoro-4-iodobutane ≥97% stabilized
1,1,1,2,2-Pentafluoro-4-iodobutane

TCA listed: T
Hazard: Toxic

Formula: C₄H₄F₅I
MW: 273.97 g/mol
Boiling Pt: 100 °C
MDL Number: MFCD00155910
CAS Number: 40723-80-6
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 100°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR